missing translation for 'onlineSavingsMsg'
Learn More

Berkshire SatPax™ 670

Product Code. 17980531 Shop All Berkshire Products
Change view
Click to view available options
Dimensions (L x W):
9 x 9 in.
Unit Size:
Pack of 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17980531 9 x 9 in. Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 17980531 Supplier Berkshire Supplier No. SPX67091112
  • ONLINE EXCLUSIVE PRICE
    266.70 €
    185.44 € / Pack of 12
    Save 81.26 €
    30% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

SatPax™ 670 pre-wetted mop wipers combine Durx™ 670 nonwoven wipers composed of a hydroentangled nonwoven blend of 55% cellulose and 45% polyester.

  • 55% cellulose and 45% polyester hydroentangled nonwoven blend
  • No chemical binders in base material
  • Pre-wetted with two saturations available: 70% IPA/ 30% DI Water to a high saturation level of 60% or 50% IPA/ 50% DI Water to a high saturation level of 70%
  • Re-sealable solvent resistant packaging
  • Wipers are c-folded and then bi-folded inwards
  • Low extractables and fiber and particle counts
  • Smooth and durable with good wet strength
  • Reduces alcohol usage and preparation/handling costs
  • Increases cleaning efficiency
  • Increases cleaning protocol consistency

Specifications

Product Type Surface Wiper
Length (English) 9 in.
Length (Metric) 23 cm
Width (English) 9 in.
Width (Metric) 23 cm
Material Cellulose
Autoclavable Not Autoclavable
Disposable Single-use
Dimensions (L x W) 9 x 9 in.
Ply 1-ply
Sterility Non-sterile
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • X12 Satpax 670 Cleanroom Presaturated Wipes, 25 per pack
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contain: isopropanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.