missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SCC1 CryoCool Heat Transfer Fluid

Product Code. 10715640 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
Explorer SpeedVac Vacuum Concentrator
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
10715640 Explorer SpeedVac Vacuum Concentrator 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10715640 Supplier Thermo Scientific™ Supplier No. SCC1
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Savant™ SCC1 CryoCool Heat Transfer Fluid is suitable for use with the Explorer SpeedVac Vacuum Concentrator.

 

  • Suitable for use with Explorer SpeedVac Vacuum Concentrator
TRUSTED_SUSTAINABILITY

Specifications

Description CryoCool™ Heat Transfer Fluid, 1L
Type Heat Transfer Fluid
For Use With (Equipment) Explorer SpeedVac Vacuum Concentrator
Model SCC1
Volume (Metric) 1 L
compliance-icons compliance-icons
Product Identifier
  • Cryocool Heat Transfer Fluid
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: OctamethyltrisiloxanOctamethylcyclotetraDodecamethylpentasilDecamethyltetrasilox
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.