missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Savant™ SpeedVac™ Medium Capacity Concentrators for Combinatorial Chemistry Applications

Product Code. 15966171 Shop All Thermo Scientific Products
Change view
Click to view available options
Model:
SpeedVac SPD130
SpeedVac SPD130DLX
Electrical Requirements:
230 V 50 Hz
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Electrical Requirements Unit Size
15966171 SpeedVac SPD130 230 V 50 Hz Each
15956171 SpeedVac SPD130DLX 230 V 50 Hz Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15966171 Supplier Thermo Scientific™ Supplier No. SPD130P1230
  • ONLINE EXCLUSIVE PRICE
    38983.95 €
    36300.00 € / Pack of 1
    Save 2683.95 €
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 29-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Evaporate strong acids, bases and aggressive organic solvents with the Thermo Scientific™ SpeedVac Modular Vacuum Concentrators. Approved for combinatorial chemistry, these concentrators incorporate an intuitive programmable user display for enhanced usability.

SPD130DLX is large capacity vacuum concentrators used with a wide variety of solvents and offer resistance to TFA, DSMO and other aggressive solvents used in combinatorial chemistry applications.   For ease of ordering, you can order this in kits.

 

SPD130DLX Features:

  • Deep aluminum standard PTFE-coated chamber
  • Safety cover lock
  • Lid stay mechanism
  • Radiant glass safety cover improves drying time by applying radiant IR heat to samples (SPD130DLX)
  • PTFE plumbing and tubing
  • Two valve manifold with built-in automatic bleeder valve
  • Time/temperature display incorporating membrane touch pad switches
  • Three preset programs by application and three user defined programs available
  • Variable drying times from 35° to 80°C in five degree increments
  • Manual/auto timed run

Solvents and Solvent Combinations:

  • Acetone
  • Chloroform
  • Ethyl acetate
  • Hexane
  • Methylene chloride

SPD 130DLX P1 Includes:

  • SpeedVac Concentrator
  • -105°C, Ultra-Low Temperature Refrigerated Vapor Trap
  • OFP400 Oil-Free ETFE/PFA 4-Headed Pump
  • 64 × 1.5mL Rotor (RH64-11)
  • Digital Vacuum Gauge
  • Chemical Trap Kit for use with disposable Cartridge
  • Tubing and Fitting Kit including 4” of clear thick walled vacuum tubing
  • Glass Condensation Flask
  • CryoCool™ Heat Transfer Fluid (1L)
TRUSTED_SUSTAINABILITY

Specifications

Model SpeedVac SPD130
Electrical Requirements 230 V 50 Hz
Certifications UL Listed to US and Canadian requirements. CE marked
Includes
  • SpeedVac Concentrator
  • -105°C, Ultra-Low Temperature Refrigerated Vapor Trap
  • OFP400 Oil-Free ETFE/PFA 4-Headed Pump
  • 64 × 1.5mL Rotor (RH64-11)
  • Digital Vacuum Gauge
  • Chemical Trap Kit for use with disposable Cartridge
  • Tubing and Fitting Kit including 4” of clear thick walled vacuum tubing
  • Glass Condensation Flask
  • CryoCool™ Heat Transfer Fluid (1L)
Type Vacuum Concentrator
compliance-icons compliance-icons
Product Identifier
  • Modular SPD130P1 Savant Speed Vac kit
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: octamethyltrisiloxane, octamethylcyclotetrasiloxane, dodecamethylpentasiloxane,decamethyltetrasilox
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.