Learn More
Thermo Scientific™ Savant™ SpeedVac™ Refrigerated Vapor Trap Series
Product Code. 15996161 Shop All Thermo Scientific Products
Change view
Click to view available options
Electrical Requirements:
230 V 50 Hz
Unit Size:
Each
Product Code. Electrical Requirements Unit Size
15996161 230 V 50 Hz Each
1 options
Product Code. 15996161 Supplier Thermo Scientific™ Supplier No. RVT450230
  • ONLINE EXCLUSIVE PRICE
    3859.20 €
    3590.00 € / Pack of 1
    Save 269.20 €
    6% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Condense and trap a wide variety of solvent vapors that can contaminate and degrade vacuum pump oil with Thermo Scientific™ Savant™ Refrigerated Vapor Traps.
Condenses and removes a wide variety of solvent vapors that can contaminate and degrade vacuum pump oil with Thermo Scientific™ Savant™ Refrigerated Vapor Traps (RVT). The RVT safely recovers the solvent and reduces emissions of hazardous chemicals in the environment.
Solvents are removed from samples during vacuum evaporation with a SpeedVac™ Concentrator which are then collected in a CFC-free Refrigerated Vapor Trap (RVT).   Solvent vapors pass into a Glass Condensation Flask and condense on its walls. This prevents the vapors from reaching and potentially damaging the oil-sealed vacuum pump. 
  • Rapid cooling time minimizes waiting period
  • Easy to remove glass flask for quick solvent changes
  • Rapidly cools to operating temperature
  • Easy to use glass condensation flask allows for the collection of liquid into a self-contained jar for easy draining
  • Sealed refrigeration system eliminates consumable coolants such as dry ice and acetone
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance UL Listed to US and Canadian requirements. CE marked
Description Refrigerated Vapor Cold Trap RVT450
Type Refrigerated Vapor Trap
Electrical Requirements 230 V 50 Hz