missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Savant™ SpeedVac™ Rotors for Large Capacity Concentrators

Product Code. 12490941 Shop All Thermo Scientific Products
Change view
Click to view available options
Holds:
1.5 mL Microtube, 12 x 75 mm, 13 x 100 mm Glass and Plastic Tubes
12 Deep-well Plates
192 Vials
30 x 15 mL Conical Centrifuge Tubes
52 x 15 mL Conical
8 Microwell Plates
Flasks
Unit Size:
Each
Pack of 2
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Holds Unit Size
12490941 192 Vials Each
11788559 30 x 15 mL Conical Centrifuge Tubes Each
11758629 52 x 15 mL Conical Each
11758559 1.5 mL Microtube, 12 x 75 mm, 13 x 100 mm Glass and Plastic Tubes Each
12410941 Flasks Pack of 2
11778569 8 Microwell Plates Each
11708569 12 Deep-well Plates Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 12490941 Supplier Thermo Scientific™ Supplier No. RH19215
  • ONLINE EXCLUSIVE PRICE
    2020.05 €
    1880.00 € / Pack of 1
    Save 140.05 €
    6% Off
Estimated Shipment: 03-03-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

SpeedVac Rotors for Large Capacity Concentrators for use with Savant SpeedVac Concentrators.

Circular Rotors:

  • Easy to use
  • Multi-size tube holes accommodate different sample sizes
  • Require no additional accessories
  • Fit directly into your concentrator
TRUSTED_SUSTAINABILITY

Specifications

Description Vials: 192 (1 dram 15 x 45mm, 4mL), Large capacity models
Type Rotor
For Use With (Equipment) SPD210, SPD2030, SPD300DDA (requires RAD200)
Holds 192 Vials
Model RH192-15
Rotor System Traditional
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.