missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Saxagliptin hydrochloride
GREENER_CHOICE

Product Code. 18783459
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18783459

Brand: Tocris Bioscience™ 6507/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

DPPIV/CD26 Inhibitors

Saxagliptin hydrochloride is a high affinity DPP-IV inhibitor (Ki = 0.6 nM). Inhibits plasma DPP-IV (87%). Elicits enhanced glucose clearance in Zuckerfa/fa rats.
TRUSTED_SUSTAINABILITY

Specifications

CAS 709031-78-7
Synonym (1S,3S,5S)-2-[(2S)-Amino(3-hydroxytricyclo[3.3.1.13,7]dec-1-yl)acetyl]-2-azabicyclo[3.1.0]hexane-3-carbonitrile hydrochloride
Quantity 50 mg
Molecular Formula C18H25N3O2.HCl
Formula Weight 351.87
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.