missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SB 202190

Product Code. 30001220
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001220 10 mg 10mg
18054982 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001220 Supplier R&D Systems™ Supplier No. 1264/10
  • ONLINE EXCLUSIVE PRICE
    294.00 €
    277.20 € / 10mg
    Save 16.80 €
    5% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective inhibitor of p38 MAPK

SB 202190 is a highly selective, potent and cell-permeable inhibitor of p38 MAP kinase. Binds within the ATP pocket of the active kinase (Kd = 38 nM, as measured in recombinant human p38), and selectively inhibits the p38α and β isoforms (IC50 = 50 and 100 nM at SAPK2a/p38 and SAPK2b/p38β2 respectively). Promotes stability of naive human pluripotent stem cells in culture. Inhibits Yamanaka factor reprogramming of human fibroblasts to iPSCs. SB 202190 improves the self-renewal ability of neuronal stem cells from NPC1-deficient mice.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152121-30-7
Molecular Formula C20H14FN3O

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 331.35
Purity >99%
Molecular Weight (g/mol) 331.35
Chemical Name or Material SB 202190
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.