missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SB 203580

Product Code. 30001347
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001347 1 mg 1mg
30000722 10 mg 10mg
18006042 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30001347 Supplier R&D Systems™ Supplier No. 1202/1
  • ONLINE EXCLUSIVE PRICE
    189.27 €
    170.10 € / 1mg
    Save 19.17 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective inhibitor of p38 MAPK

SB 203580 is a selective inhibitor of p38 MAPK (IC50 values are 50 and 500 nM for SAPK2a/p38 and SAPK2b/p38β2 respectively). Displays 100-500-fold selectivity over LCK, GSK-3β and PKBα. Shown to inhibit IL-2-induced T cell proliferation, cyclooxygenase-1 and -2, and thromboxane synthase. Enhances clonal growth of skin epithelial progenitor cells; stimulates neural stem cell (NSC) proliferation. SB 203580 is an essential component of medium for maintaining stem cells in naive pluripotent state and can be used to promote expansion of HSCs ex vivo.

Water-soluble Salt also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152121-47-6
Molecular Formula C21H16FN3OS

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 377.44
Purity >98%
Molecular Weight (g/mol) 377.437
Chemical Name or Material SB 203580
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.