Learn More
SB 203580 hydrochloride, Tocris Bioscience™
GREENER_CHOICE
Selective inhibitor of p38 MAPK; water-soluble
Brand:  Tocris Bioscience 1402/10
Encompass
Quantity 10mg
Product Code. 13449323
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Water-soluble salt of SB 203580 (Cat. No. 1202). Selective inhibitor of p38 mitogen-activated protein kinase (IC50 values are 50 and 500 nM for SAPK2a/p38 and SAPK2b/p38β2 respectively). Displays 100-500-fold selectivity over LCK, GSK3β and PKBα. Shown to inhibit interleukin-2-induced T cell proliferation, cyclooxygenase-1 and -2, and thromboxane synthase.
Specifications
Specifications
869185-85-3
10mg
WOSGGXINSLMASH-UHFFFAOYNA-N
413.89
SB 203580 hydrochloride
C21H17ClFN3OS
4-[5-(4-Fluorophenyl)-2-[4-(methylsulphonyl)phenyl]-1H-imidazol-4-yl]pyridine hydrochloride
413.90
>98%