missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SB 203580 hydrochloride
GREENER_CHOICE

Product Code. 18789896
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18789896 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18789896 Supplier Tocris Bioscience™ Supplier No. 1402/10
  • ONLINE EXCLUSIVE PRICE
    423.00 €
    400.05 € / 10mg
    Save 22.95 €
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective inhibitor of p38 MAPK; water-soluble

Water-soluble salt of SB 203580 (Cat. No. 1202). Selective inhibitor of p38 mitogen-activated protein kinase (IC50 values are 50 and 500 nM for SAPK2a/p38 and SAPK2b/p38β2 respectively). Displays 100-500-fold selectivity over LCK, GSK3β and PKBα. Shown to inhibit interleukin-2-induced T cell proliferation, cyclooxygenase-1 and -2, and thromboxane synthase.
TRUSTED_SUSTAINABILITY

Specifications

CAS 869185-85-3
Quantity 10 mg
Molecular Formula C21H17ClFN3OS
Formula Weight 413.89
Purity >98%
Molecular Weight (g/mol) 413.90
Chemical Name or Material SB 203580 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.