missing translation for 'onlineSavingsMsg'
Learn More

SB 203580 Hydrochloride, TRC

Product Code. 30424510
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424510 10 mg -
30414831 50 mg -
30411094 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424510 Supplier Toronto Research Chemicals Supplier No. TRCS15505810MG
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 869185-85-3
Molecular Formula C21H16FN3OS . HCl
Molecular Weight (g/mol) 413.9
Synonym 4-[5-(4-Fluorophenyl)-2-[4-(methylsulphonyl)phenyl]-1H-imidazol-4-yl]pyridine Hydrochloride (1:1), 4-[5-(4-Fluorophenyl)-2-[4-(methylsulphonyl)phenyl]-1H-imidazol-4-yl]pyridine Monohydrochloride (9CI), SB 203580 Hydrochloride
IUPAC Name 4-[4-(4-fluorophenyl)-2-(4-methylsulfinylphenyl)-1H-imidazol-5-yl]pyridine;hydrochloride
SMILES Cl.CS(=O)c1ccc(cc1)c2nc(c3ccc(F)cc3)c([nH]2)c4ccncc4

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H16FN3OS.ClH/c1-27(26)18-8-4-16(5-9-18)21-24-19(14-2-6-17(22)7-3-14)20(25-21)15-10-12-23-13-11-15;/h2-13H,1H3,(H,24,25);1H
Formula Weight 413.0765
Chemical Name or Material 4-[5-(4-Fluorophenyl)-2-[4-(methylsulphonyl)phenyl]-1H-imidazol-4-yl]pyridine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.