missing translation for 'onlineSavingsMsg'
Learn More

SB 204990, MedChemExpress

Product Code. 16479597
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479597 5 mg 5mg
16427058 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16479597 Supplier Medchem Express Supplier No. HY16450
 Restricted Item
Estimated Shipment: 15-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress SB 204990 is a potent and specific inhibitor of ATP citrate lyase (ACLY) enzyme.

SB 204990 is a potent and specific inhibitor of ATP citrate lyase (ACLY) enzyme.

Specifications

Chemical Name or Material SB 204990
CAS 154566-12-8
Color White
Purity Grade Notes Research
Molecular Formula C18H22Cl2O5
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (256.89 mM) ∣H2O : 25 mg/mL (64.22 mM; Need ultrasonic)
SMILES O=C(O)C[C@]1(O)C(O[C@H](CCCCCCC2=CC=C(Cl)C=C2Cl)C1)=O
Molecular Weight (g/mol) 389.27
Formula Weight 389.27
Percent Purity 95.35%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.