missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals SB 225002

Product Code. 15496869
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15496869 10 mg 10mg
15486869 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15496869 Supplier Thermo Scientific Chemicals Supplier No. J65387.MA
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

A potent, selective and competitive antagonist of the G-protein coupled receptor CXCR2

SB 225002 is a selective non-peptide CXCR2 inhibitor which suppresses IL-8 induced neutrophil chemotaxis and margination. In addition, SB 225002 reduces MPO activity, IL-1β, MIP-2, and keratinocyte-derived chemokine (KC) levels. Further studies suggest that SB 225002 decreases the expression of vascular endothelial growth factor, inducible NOS (NO synthase), and Cox-2 (cyclooxygenase-2 proteins) in colon tissue.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 182498-32-4
Molecular Formula C13H10BrN3O4, C13H10BrN3O4
Synonym 1-(2-Bromophenyl)-3-(2-hydroxy-4-nitrophenyl)urea

Specifications

Quantity 10 mg
Description Yellow
Content And Storage -30°C to -10°C
For Use With (Application) A potent, selective, and competitive antagonist of the G-protein coupled receptor CXCR2
Formula Weight 352.10g/mol
Sensitivity Moisture Sensitive
Solubility Soluble to 100mM in DMSO and to 50mM in ethanol
Product Type Signal Transduction Reagent
Form Solid
InChI Key MQBZVUNNWUIPMK-UHFFFAOYSA-N
SMILES OC1=C(NC(=O)NC2=CC=CC=C2Br)C=CC(=C1)[N+]([O-])=O
IUPAC Name 3-(2-bromophenyl)-1-(2-hydroxy-4-nitrophenyl)urea
Molecular Weight (g/mol) 352.14
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.