missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SB 225002

Product Code. 18084762
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18084762 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18084762 Supplier R&D Systems™ Supplier No. 2725/50
  • ONLINE EXCLUSIVE PRICE
    1330.04 €
    1190.70 € / 50mg
    Save 139.34 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective CXCR2 antagonist

Potent and selective CXCR2 chemokine receptor antagonist (IC50 = 22 nM) that displays > 150-fold selectivity over CXCR1 receptors. Causes inhibition of IL-8 and GROα-mediated calcium mobilization in HL60 cells (IC50 values are 8 and 10 nM respectively). Prevents IL-8-induced neutrophil chemotaxis in vitro and sequestration in vivo. Inhibits HIV replication in lymphocytes and macrophages.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 182498-32-4
Molecular Formula C13H10BrN3O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 352.14
Purity >99%
Molecular Weight (g/mol) 352.14
Chemical Name or Material SB 225002
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.