missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SB 258585 hydrochloride
GREENER_CHOICE

Product Code. 30002124
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30002124 50 mg 50mg
30001289 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30002124 Supplier Tocris Bioscience™ Supplier No. 1961/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent, selective 5-HT6 antagonist

Potent and selective 5-HT6 receptor antagonist (pKi = 8.6); displays > 160-fold selectivity over other 5-HT receptor subtypes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1216468-02-8
Molecular Formula C18H23ClIN3O3S

Specifications

Quantity 50 mg
Formula Weight 523.82
Purity >99%
Molecular Weight (g/mol) 523.814
Chemical Name or Material SB 258585 hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.