missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SB 258585 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent and selective 5-HT6 receptor antagonist (pKi = 8.6); displays > 160-fold selectivity over other 5-HT receptor subtypes.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 523.82 |
| Purity | >99% |
| Molecular Weight (g/mol) | 523.814 |
| Chemical Name or Material | SB 258585 hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction