missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SB 4

Product Code. 18759586
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18759586 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18759586 Supplier R&D Systems™ Supplier No. 6881/50
  • ONLINE EXCLUSIVE PRICE
    198.49 €
    177.45 € / 50mg
    Save 21.04 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent BMP4 agonist

SB 4 is a potent BMP4 agonist (EC50 = 74 nM). Activates canonical BMP signaling and increases SMAD-1/5/9 phosphorylation, with no effect on p-SMAD-2, p-SMAD-3 or p-TAK1. Bypasses noggin-induced inhibition of type I BMP receptors.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[[(4-Bromophenyl)methyl]thio]benzoxazole
CAS 100874-08-6
Quantity 50 mg
Product Line Tocris
Molecular Formula C14H10BrNOS
Formula Weight 320.2
Product Type Agonist
Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.