missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ SB 431542

Product Code. 18795776
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18795776 10 mg 10mg
18799876 1 mg 1mg
18064922 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18795776 Supplier R&D Systems™ Supplier No. 1614/10
  • ONLINE EXCLUSIVE PRICE
    468.58 €
    417.90 € / 10mg
    Save 50.68 €
    10% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent, selective inhibitor of TGF-βRI, ALK4 and ALK7

SB 431542 is a potent and selective inhibitor of the transforming growth factor-β (TGF-β) type I receptor/ALK5 (IC50 = 94 nM), and its relatives ALK4 and ALK7. Suppresses TGF-β-induced proliferation of human osteosarcoma cells. Replaces SOX2 in reprogramming of fibroblasts into iPSCs. Stimulates proliferation, differentiation and sheet formation of ESC-derived endothelial cells. Inhibits TGF-β-induced EMT, migration, invasion and VEGF secretion in several human cancer cell lines.

SB 431542 synthesized to cGMP guidelines also available.

For more information about how SB 431542 may be used, see our protocols: 3D Culture of Lung Alveolar Cells, Differentiation of Hematopoietic Progenitors from hPSCs, Generating Midbrain Dopaminergic Neurons from hPSCs, Generating Vascular Organoids, Accelerated Induction of Cortical Neurons from hiPSCs .

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material SB 431542
CAS 301836-41-9
Synonym 4-4-2h-1,3-benzodioxol-5-yl-5-pyridin-2-yl-1h-imidazol-2-yl benzamide
Quantity 10 mg
Product Line Tocris
Molecular Formula C22H16N4O3
Formula Weight 384.39
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.