missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SB 699551
GREENER_CHOICE

Product Code. 18004792
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18004792 10 mg 10mg
18065522 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18004792 Supplier Tocris Bioscience™ Supplier No. 3188/10
  • ONLINE EXCLUSIVE PRICE
    316.00 €
    298.20 € / 10mg
    Save 17.80 €
    5% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Selective 5-ht5a antagonist

Selective 5-ht5a receptor antagonist (pKi values are 8.3, < 6.0, < 6.0, < 6.0, < 5.5 and < 5.5 for 5-ht5a, 5-HT1B/D, 5-HT2A, 5-HT2C, 5-HT1A and 5-HT7 receptors respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 864741-95-7
Molecular Formula C34H47Cl2N3O

Specifications

Quantity 10 mg
Formula Weight 584.66
Purity >99%
Molecular Weight (g/mol) 584.67
Chemical Name or Material SB 699551
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.