missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SB 699551

Description
Selective 5-ht5a receptor antagonist (pKi values are 8.3, < 6.0, < 6.0, < 6.0, < 5.5 and < 5.5 for 5-ht5a, 5-HT1B/D, 5-HT2A, 5-HT2C, 5-HT1A and 5-HT7 receptors respectively).
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 584.66 |
| Purity | >99% |
| Molecular Weight (g/mol) | 584.67 |
| Chemical Name or Material | SB 699551 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?