missing translation for 'onlineSavingsMsg'
Learn More

SC 514, Tocris Bioscience™

Product Code. 18085512
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18085512

Brand: Tocris Bioscience 3318/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

IKKβ inhibitor; attenuates NF-κB-induced gene expression

Orally active, ATP-competitive IKKβ inhibitor (IC50 = 3 - 12 μM) that displays > 10-fold selectivity over 28 other kinases including JNK, p38, MK2 and ERK. Attenuates NF-κB-induced gene expression of IL-6, IL-8 and COX-2 in synovial fibroblasts (IC50 values are 20, 20 and 8 μM respectively). Reduces iNOS induction and exhibits anti-inflammatory activity in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 354812-17-2
Molecular Formula C9H8N2OS2
Molecular Weight (g/mol) 224.296
InChI Key BMUACLADCKCNKZ-UHFFFAOYSA-N
Synonym 3-amino-5-thiophen-3-yl thiophene-2-carboxamide, 4-amino-2,3'-bithiophene-5-carboxamide, ikk-2 inhibitor, sc-514, insolution ikk-2 inhibitor, sc-514, 5-thien-3-yl-3-aminothiophene-2-carboxamide, curator_000007, 4-amino-2,3-bithiophene-5-carboxamide, biomolki_000076, biomolki2_000080, d0t6va
PubChem CID 2807869
IUPAC Name 3-amino-5-thiophen-3-ylthiophene-2-carboxamide
SMILES C1=CSC=C1C2=CC(=C(S2)C(=O)N)N

Specifications

Quantity 10 mg
Formula Weight 224.3
Percent Purity >98%
Chemical Name or Material SC 514
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.