missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sca-1/Ly6 Rat anti-Mouse, Alexa Fluor 750, Clone: 177228, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB1226S-100UG
This item is not returnable.
View return policy
Description
Sca-1/Ly6 Monoclonal specifically detects Sca-1/Ly6 in Mouse samples. It is validated for Flow Cytometry.Specifications
| Sca-1/Ly6 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| CAA28351 | |
| LY6A | |
| Mouse myeloma cell line NS0-derived recombinant mouse Sca-1/Ly6 C-terminally truncated Ly-6E allele Leu27-Gly119 Accession # CAA28351 | |
| RUO | |
| 58530 | |
| Mouse | |
| IgG2a |
| Flow Cytometry | |
| 177228 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| C6orf23, G6D, Ly6, Ly-6, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25, Sca1 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse Sca-1/Ly6 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction