missing translation for 'onlineSavingsMsg'
Learn More

Scandium foil, 0.025mm (0.001in) thick, 99% (REO)

Product Code. 11301799
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
Unit Size:
25mm
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11301799 25 x 25 mm 25mm
11440280 50 x 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11301799 Supplier Thermo Scientific Alfa Aesar Supplier No. 012426.FF
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7440-20-2 | Sc | 44.96 g/mol

It finds applications in the aerospace industry. Another use of Scandium is in aluminum-scandium alloys for sports equipment, such as bicycle frames, lacrosse sticks, and baseball bats.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-20-2
Molecular Formula Sc
Molecular Weight (g/mol) 44.96
MDL Number MFCD00016323
InChI Key SIXSYDAISGFNSX-UHFFFAOYSA-N
Synonym unii-yuj4u1ew7r, yuj4u1ew7r, escandio, skandium, atom, powder, ingot, nitrate solution, standard for aas, scandium, lump, vacuum remelted
PubChem CID 23952
ChEBI CHEBI:33330
IUPAC Name scandium
SMILES [Sc]

Specifications

Color Gray
Quantity 25 x 25 mm
Odor Odorless
Weight ≈0.05g/25x25mm
Form Foil
UN Number UN3178
Solubility Information Slowly dissolve in dilute acids.
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Scandium foil, 0.025mm (0.001 in.) thick

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.