missing translation for 'onlineSavingsMsg'
Learn More

Scandium(III) acetate hydrate, 99.9% (metals basis)

Product Code. 11463817
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463817 1 g 1g
11473817 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463817 Supplier Thermo Scientific Alfa Aesar Supplier No. A18271.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 304675-64-7 | C6H9O6Sc | 222.09 g/mol

Scandium(III) acetate hydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Scandium(III) acetate hydrate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304675-64-7
Molecular Formula C6H9O6Sc
Molecular Weight (g/mol) 222.09
MDL Number MFCD00150122
InChI Key DBTMQFKUVICLQN-UHFFFAOYSA-K
IUPAC Name scandium(3+) triacetate
SMILES [Sc+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Linear Formula (CH3CO2)3Sc·xH2O
Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 222.10 (Anhydrous)
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Scandium(III) acetate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.