Learn More
Merck Scepter Cell Counter Sensors
Product Code. 15421700
Change view
Click to view available options
Quantity:
250/Pk
Unit Size:
Pack of 250
Product Code. Quantity Unit Size
15421700 250/Pk Pack of 250
1 options
Product Code. 15421700 Supplier Merck Supplier No. PHCC60250
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Ver productos alternativos
missing translation for 'validMainframeMsg'
Este artículo no se puede devolver. Vea la política de devoluciones
Scepter Cell Counter Sensors are engineered with a micro-fabricated, cell-sensing zone that enables discrimination by cell size and cell volume down to sub-micron and sub-picoliter resolution.
  • Coupled with precision liquid handling channels and electronics, the Scepter sensor accurately and reliably provides cell population statistics.
  • The Scepter 2.0 handheld automated cell counter provides a fast and convenient method for counting cells and particles and is compatible with both the 40 and 60uM sensor. The system employs the Coulter principle in a miniaturized, handheld, format enabling rapid cell counting; what used to take 10 minutes now takes less than 30 seconds. The user prepares a dilution of the cell culture of interest and uses the Scepter cell counter to aspirate a sample of this dilution into the Scepter sensor.
  • The Scepter cell counter’s screen displays:
    • Cell concentration 
    • Average cell size 
    • Average cell volume 
    • Histogram of size or volume distribution
    • This system is intended for research use only and has been tested with cell types representative of those in use today
  • 60 μm sensors designed to count particles between 6 μm and 36 μm
  • Operating Range: 10,000 particles/mL–500,000 particles/mL
  • Partical Size: 6 μm–36 μm
  • Cell Size range: 8.0–25.0 μm
TRUSTED_SUSTAINABILITY
Especificaciones
Includes 50 μL sample volume
Quantity 250/Pk
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.