missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Rat IgM (H+L) Secondary Antibody, DyLight 550, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-68494R
This item is not returnable.
View return policy
Description
IgM (H+L) Polyclonal antibody specifically detects IgM (H+L) in Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA
Specifications
| IgM (H+L) | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rat IgM | |
| 0.5 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA | |
| DyLight 550 | |
| Goat | |
| Affinity purified | |
| RUO | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction