missing translation for 'onlineSavingsMsg'
Learn More

SCH 28080, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18065342
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18065342

Brand: Tocris Bioscience 1690/10

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

H+,K+-ATPase inhibitor

Potent inhibitor of H+,K+-ATPase (IC50 = 20 nM); binds to the K+ recognition site and is competitive with respect to K+. Inhibits gastric acid secretion in vitro and in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76081-98-6
Molecular Formula C17H15N3O
Molecular Weight (g/mol) 277.33
MDL Number MFCD00834620
InChI Key PYKJFEPAUKAXNN-UHFFFAOYSA-N
Synonym schering compound 28080, 2-methyl-8-phenylmethoxy imidazo 1,2-a pyridine-3-acetonitrile, chembl47529, 2-methyl-8-phenylmethoxy imidazo-1-2-a pyrine-3-acetonitrile, 2-8-benzyloxy-2-methylimidazo 1,2-a pyridin-3-yl acetonitrile, imidazo 1,2-a pyridine-3-acetonitrile, 2-methyl-8-phenylmethoxy, tocris-1690, 8-benzyloxy-2-methylimidazo 1,2-a pyridin-3-yl acetonitrile, lopac-s-4443
PubChem CID 108137
IUPAC Name 2-[8-(benzyloxy)-2-methylimidazo[1,2-a]pyridin-3-yl]acetonitrile
SMILES CC1=C(CC#N)N2C=CC=C(OCC3=CC=CC=C3)C2=N1

Specifications

Quantity 10 mg
Formula Weight 277.33
Percent Purity >99%
Chemical Name or Material SCH 28080
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.