Learn More
Thermo Scientific™ NanoDrop™ Eight SciVault Compliance Software
Product Code. 31921083 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
NanoDrop Eight Spectrophotometers
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31921083 NanoDrop Eight Spectrophotometers Each
1 options
Product Code. 31921083 Supplier Thermo Scientific™ Supplier No. 840371100
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Confidently secure your laboratory data and more easily achieve compliance with US FDA 21 CFR Part 11 with Thermo Scientific™ SciVault Software for use with the NanoDrop™ Eight Microvolume Spectrophotometer. The software helps to facilitate compliant results in regulated environments with advanced tools such as user account access control, electronic signatures, audit trails, automated data exports, and security policies.
Confidently secure your laboratory data and more easily achieve compliance with US FDA 21 CFR Part 11 with Thermo Scientific™ SciVault Software for use with the NanoDrop™ Eight Microvolume Spectrophotometer. The software helps to facilitate compliant results in regulated environments with advanced tools such as user account access control, electronic signatures, audit trails, automated data exports, and security policies. SciVault software integrates directly into the NanoDrop Eight Software for streamlined operation.
Thermo Scientific™ SciVault™ Software helps your lab stay compliant with US FDA data regulations every step of the way.
  • Streamline management –SciVault Software integrates into NanoDrop Eight Software user interface
  • Comply with US FDA 21 CFR Part 11 Regulation – Control user account access, apply digital signatures, and review audit logs
  • Network multiple instruments – Install SciVault Software on a central computer to control privileges and view audit logs across multiple NanoDrop Eight Instruments in different labs
  • Validate your systems – IQ/OQ documents and services available to confirm your instrument is running within manufacturer’s specifications
Specifications
Description SciVault compliance software for NanoDrop Eight Spectophotometers
For Use With (Equipment) NanoDrop Eight Spectrophotometers
Includes Software is designed to integrate into the NanoDrop Eight PC software user interface. Software enable users to run the NanoDrop Eight instrument in a lab that needs to comply with US FDA 21 CFR Part 11. Includes: USB flash drive containing the SciVault software for the NanoDrop Eight Spectrophotometer