missing translation for 'onlineSavingsMsg'
Learn More

Sclareolide, TRC

Product Code. 30427288
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30427288 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427288 Supplier Toronto Research Chemicals Supplier No. TRCS1999901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sclareolide
CAS 564-20-5
Molecular Formula C16 H26 O2
Quantity 1 g
Synonym (3aR)-(+)-Sclareolide, (+)-12-Norambreinolide, Decahydrotetramethylnaphthofuranone
InChI Formula InChI=1S/C16H26O2/c1-14(2)7-5-8-15(3)11(14)6-9-16(4)12(15)10-13(17)18-16/h11-12H,5-10H2,1-4H3/t11-,12+,15-,16+/m0/s1
SMILES CC1(C)CCC[C@@]2(C)[C@H]1CC[C@@]3(C)OC(=O)C[C@H]23
IUPAC Name (3aR,5aS,9aS,9bR)-3a,6,6,9a-tetramethyl-1,4,5,5a,7,8,9,9b-octahydrobenzo[e]benzofuran-2-one
Molecular Weight (g/mol) 250.38
Formula Weight 250.1933
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.