missing translation for 'onlineSavingsMsg'
Learn More

Scopolamine hydrobromide trihydrate, 99%

Product Code. 10117943 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10117943 1 g Glass bottle 1g
10605222 5 g Glass bottle 5g
10289593 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10117943 Supplier Thermo Scientific Acros Supplier No. 161750010
Curiosity Goes Beyond
Use promo code 27966
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Scopolamine hydrobromide trihydrate, CAS # 6533-68-2, is the trihydrate form of scopolamine hydrobromide that shows competitive anti-muscarinic activity by blocking the actions of endogenous acetylcholine or exogenous agonists. It also shows anti-spasmodic and anti-emetic activities. | CAS: 6533-68-2 | C17H28BrNO7 | 438.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Information

• Scopolamine hydrobromide trihydrate is a salt form of scopolamine, a tropane alkaloid derived from plants of the nightshade family (Solanaceae), mainly Hyoscyamus niger and Atropa belladonna
• Scopolamine hydrobromide trihydrate, demonstrates competitive anti-muscarinic activity blocking the actions of endogenous acetylcholine or exogenous agonists
• It also shows anti-spasmodic and anti-emetic activities

Applications

• It has been described that scopolamine inhibits muscarinic acetylcholine receptors (mAChRs) with some selectivity for M1 mAChRs
• It is structurally similar to acetylcholine and can antagonize acetylcholine activity mediated by muscarinic receptors located on structures innervated by postganglionic cholinergic nerves and on smooth muscles (that respond to acetylcholine but lack cholinergic innervation)
• Scopolamine hydrobromide trihydrate can induce amnesia in laboratory animals and so it is frequently used in animal models of memory deficit
• Scopolamine hydrobromide trihydrate is used to induce cognitive deficits in animal models of Alzheimer’s disease
• Scopolamine, has been employed in several studies to understand, identify, and characterize biological targets for Alzheimer's disease

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6533-68-2
Molecular Formula C17H28BrNO7
Molecular Weight (g/mol) 438.32
MDL Number MFCD00150066
InChI Key LACQPOBCQQPVIT-FCFGRTKISA-N
Synonym hyoscine hydrobromide trihydrate
PubChem CID 131632304
IUPAC Name hydrogen (1S,2R,4S,5S)-9-methyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-7-yl (2S)-3-hydroxy-2-phenylpropanoate trihydrate bromide
SMILES [H+].O.O.O.[Br-].CN1[C@H]2CC(C[C@H]1[C@@H]1O[C@H]21)OC(=O)[C@H](CO)C1=CC=CC=C1

Specifications

Melting Point 194.0°C to 197.0°C
Color White or Colorless
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Beilstein 27, 102
Merck Index 14, 8406
Specific Rotation Condition −27° (20°C c=5,H2O)
Specific Rotation − 27.00
Solubility Information Solubility in water: 700g/L water (20°C). Other solubilities: 1g/20 mL alcohol,slightly soluble in chloroform,practically insoluble in ether
Formula Weight 438.3
Percent Purity 99%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Scopolamine hydrobromide trihydrate, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Scopolamine hydrobromide trihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.