missing translation for 'onlineSavingsMsg'
Learn More

Scopoletin, MedChemExpress

Product Code. 16479056
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16479056 100 mg 100mg
16454817 50 mg 50mg
16431506 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16479056 Supplier Medchem Express Supplier No. HYN0342
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Scopoletin is an inhibitor of acetylcholinesterase (AChE).

Scopoletin is an inhibitor of acetylcholinesterase (AChE).

Specifications

Chemical Name or Material Scopoletin
CAS 92-61-5
Color Light Nude
Purity Grade Notes Research
Molecular Formula C10H8O4
Quantity 100 mg
Synonym Gelseminic acid Chrysatropic acid
Solubility Information DMSO : 62.5 mg/mL (325.23 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble)
SMILES O=C1C=CC2=CC(OC)=C(O)C=C2O1
Molecular Weight (g/mol) 192.17
Formula Weight 192.17
Percent Purity 99.19%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.