missing translation for 'onlineSavingsMsg'
Learn More

Scopoletin, TRC

Product Code. 30424106
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30424106 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30424106 Supplier Toronto Research Chemicals Supplier No. TRCS200500100MG
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Scopoletin
CAS 92-61-5
Purity Grade Notes HPLC
Molecular Formula C10 H8 O4
Quantity 100 mg
Synonym Coumarin','7-hydroxy-6-methoxy- (8CI), 7-Hydroxy-6-methoxy-2H-1-benzopyran-2-one, 6-Methoxy-7-hydroxycoumarin, 6-Methylesculetin, 6-O-Methylesculetin, 7-Hydroxy-6-methoxycoumarin, Buxuletin, Chrysatropic acid, Escopoletin, Esculetin 6-methyl ether, Gelseminic acid, Murrayetin, NSC 405647, Scopoletin, Scopoletine, Scopoletol, β-Methylesculetin
InChI Formula InChI=1S/C10H8O4/c1-13-9-4-6-2-3-10(12)14-8(6)5-7(9)11/h2-5,11H,1H3
SMILES COc1cc2C=CC(=O)Oc2cc1O
IUPAC Name 7-hydroxy-6-methoxychromen-2-one
Molecular Weight (g/mol) 192.17
Formula Weight 192.0423
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.