missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ Scorpio™ 8-354 Series Green Neoprene Immersion Gloves

Product Code. 12669791
Change view
Click to view available options
Size:
8
9
10
Unit Size:
12 pairs
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12669791 10 12 pairs
12679791 8 12 pairs
12055550 9 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12669791 Supplier Ansell™ Supplier No. 8354100
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Neoprene dipped coating on interlock knit liner for better grip on wet and slippery materials.

  • Flexible
  • Textured finish
  • 100% liquidproof
  • Soft stretchy knit liner with no seams in working area
  • Comfortable to wear, cushions the hand, absorbs perspiration and prevents chafing
  • Good wet grip. Safer handling means increased productivity
  • Easier to manipulate, less tiring to the hand and easier to get on and off
  • Neoprene coating
  • Wide ranging chemical protection
  • Certified FDA21 CFR 177-2600

Applications:

  • Cleaning furnaces, distillation pumps, valves or lines and crackers BTX process
  • Equipment repair and maintenance
  • Filling, blending and charging of raw materials
  • Loading and unloading process equipment
  • Opening and draining pumps valves or lines
  • Opening furnaces draining pumps valves or lines and crackers BTX process
  • Transferring liquids and solids
  • Supervision of running operation
  • Warehousing of raw materials
  • Transferring liquids and solids between vessels and tanks and process equipment
TRUSTED_SUSTAINABILITY

Specifications

Back Material Neoprene
Certifications/Compliance ANSI, EN 388 : 3121, EN 374
Cuff Material Neoprene
Coating Material Neoprene
Color Green
Exterior Finish Rough
Hand Specific
Laboratory/Cleanroom Use Yes
Latex Free Yes
Material Neoprene
Product Type Immersion Gloves
Size 10
Glove Cuff Style Gauntlet
Liner Material Interlock Knit Cotton
Lining Knit Lining
Palm Type Fully coated-palm
Quantity 72 Pairs
Resistant To Chemicals
Series 8-354
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.