Learn More
3M™ Scotch™ Performance Box Sealing Tape 3739
Product Code. 17811700
Change view
Click to view available options
Color:
Brown
Transparent
Unit Size:
Pack of 36
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
17811700 Brown Pack of 36
17821700 Transparent Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17811700 Supplier 3M™ Supplier No. 7000095400
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Mainly used for single strip closure of fiberboard boxes, resealing, and packaging of heavy weight goods. Performs well throughout all temperatures typically encountered by packaged products in shipping and storage environments.
  • Reseal and package heavy weights of valuable goods
  • Reliable box sealing tape for medium- to heavy-weight packaging applications
  • High initial tack and shear adhesion to fiberboard
  • Use for single strip closure of fiberboard boxes
  • Performs well at all temperatures typically encountered by packaged products in shipping and storage environments
  • Quality tape for closing boxes
  • Tape resists splitting when subjected to rough handling and absorbs impact shock
TRUSTED_SUSTAINABILITY
Specifications
Adhesive Type Hot-melt
Color Brown
Format Tape
Product Type Tape
Dimensions (L x W) 66 m x 50 mm
Length (English) 72.18 yd.
Width (English) 2 in.