missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SCOTfluor glucose probe 510
GREENER_CHOICE

Product Code. 18719006
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18719006 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18719006 Supplier Tocris Bioscience™ Supplier No. 7447/2
  • ONLINE EXCLUSIVE PRICE
    271.00 €
    256.20 € / 2mg
    Save 14.80 €
    5% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Fluorescent glucose probe

SCOTfluor glucose probe 510 is a fluorescent probe for visualizing glucose uptake in vivo. It can be multiplexed with BFP and GFP. Excitation and emission maxima (λ) are 490 and 605 nm, respectively.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Deoxy-2-[(7-nitro-2,1,3-benzoselenadiazol-4-yl)amino]-D-glucose
CAS 2490298-72-9
Quantity 2 mg
Target Fluorescent Transporter Probes
Molecular Formula C12H14N4O7Se
Purity 0.98
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.