missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble Chase™ Screw Caps for Disposable Glass Tubes

Product Code. 11587133
Change view
Click to view available options
Closure Size:
15-415
18-415
Unit Size:
Pack of 1000
Pack of 500
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size unitSize
11587133 15-415 Pack of 1000
11597133 18-415 Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11587133 Supplier DWK Life Sciences Supplier No. 73805B15415
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Linerless, one-piece closures with inner sealing ring

  • Polypropylene
  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ASTM E982
Diameter (Metric) Outer 16 mm
Closure Size 15-415
Product Type Screw Cap
Autoclavable Autoclavable
Description Locking Screw Cap
Material Polypropylene
Color White
Quantity 1000 Pack
For Use With (Equipment) Culture Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.