missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Kimble Chase™ Screw Caps for Disposable Glass Tubes
Description
- Polypropylene
- Autoclavable
Specifications
Specifications
| Certifications/Compliance | ASTM E982 |
| Closure Size | 18-415 |
| Product Type | Screw Cap |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| Color | Natural |
| For Use With (Equipment) | Disposable Glass Tubes |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?