missing translation for 'onlineSavingsMsg'
Learn More

Labcon Screw Caps with O-Rings for SuperClear™ Microtubes
GREENER_CHOICE

Product Code. 16654102
Click to view available options
Color:
Assorted
Blue
Green
Natural
Orange
Purple
Red
White
Yellow
Unit Size:
Pack of 500
Pack of 5000
This item is not returnable. View return policy

Product Code. 16654102

Brand: Labcon 36008720009

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

With ethylene propylene O-ring

  • Microtube cap
  • With ethylene propylene O-ring
  • Endotoxin Free (non-pyrogenic)
  • Polyethylene USP Class VI resin
  • Made in USA

Specifications

Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Liner Material Ethylene Propylene O-ring
Pre-sterilized No
Product Type Microtube Cap
Autoclavable Autoclavable
Material Polyethylene
Color Yellow
For Use With (Application) Centrifugation
Packaging 500/Bag, 10 Bags/Cs.
Quantity 500/Bag
Product Line SuperClear
Clean Claims Endotoxin Free (non-pyrogenic)
Sterility Non-sterile
For Use With (Equipment) For Cell Culture and General Laboratory Use
Disposable Single-use
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.