missing translation for 'onlineSavingsMsg'
Learn More

Labcon Screw Caps with O-Rings for SuperClear™ Microtubes
GREENER_CHOICE

Product Code. 16694102
Change view
Click to view available options
Color:
Assorted
Blue
Green
Natural
Orange
Purple
Red
White
Yellow
Unit Size:
Pack of 500
Pack of 5000
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
16694102 Blue Pack of 500
16634102 Natural Pack of 500
16644102 White Pack of 500
16654102 Yellow Pack of 500
16664102 Orange Pack of 500
16674102 Red Pack of 500
16684102 Purple Pack of 500
16604112 Green Pack of 500
17577606 Assorted Pack of 5000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 16694102 Supplier Labcon Supplier No. 36008770009
Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

With ethylene propylene O-ring

  • Microtube cap
  • With ethylene propylene O-ring
  • Endotoxin Free (non-pyrogenic)
  • Polyethylene USP Class VI resin
  • Made in USA

Specifications

Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Liner Material Ethylene Propylene O-ring
Pre-sterilized No
Product Type Microtube Cap
Autoclavable Autoclavable
Material Polyethylene
Color Blue
For Use With (Application) Centrifugation
Packaging 500/Bag, 10 Bags/Cs.
Quantity 500/Bag
Product Line SuperClear
Clean Claims Endotoxin Free (non-pyrogenic)
Sterility Non-sterile
For Use With (Equipment) Microcentrifuge Tube
Disposable Single-use
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.