missing translation for 'onlineSavingsMsg'
Learn More

SCAT Screw Caps for Schott Duran™ Laboratory Bottles

Product Code. 15347439 Shop All SCAT Europe Products
Change view
Click to view available options
Color:
Blue
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15347439 Blue Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15347439 Supplier SCAT Supplier No. 199011
  • ONLINE EXCLUSIVE PRICE
    7.65 €
    7.00 € / Pack of 1
    Save 0.65 €
    8% Off
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Screw caps for borosilicate glass laboratory bottles.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

Specifications

Closure Size GL45
Product Type Screw Cap
Autoclavable Not Autoclavable
Material Polypropylene
Color Blue
Sterility Non-sterile
For Use With (Equipment) Schott Duran™ Bottles
Thread Size GL45
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.