missing translation for 'onlineSavingsMsg'
Learn More

Globe Scientific Screwcaps for Storage Tubes

Product Code. 31922657 Shop All Globe Scientific Products
Change view
Click to view available options
Color:
Natural
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
31922657 Natural Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31922657 Supplier Globe Scientific Supplier No. 6136
Estimated Shipment: 11-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for use with the Globe Scientific sample tubes. Globe Scientific Screwcaps for Storage Tubes are made of polypropylene, providing the same coefficient of expansion as the tubes they cap. This, along with a polypropylene inner lip, ensures a leak-proof seal even at changing temperatures.

  • For use with the Globe Scientific sample tubes (21-200-102)
  • Polypropylene construction provides the same coefficient of expansion as the tubes they fit
  • Polypropylene inner lip ensures a leakproof seal even at changing temperatures
  • Features a long skirt and a super-fast thread design that allows it to be removed or sealed with a single turn

Specifica

Product Type Screw Cap with O-ring
Autoclavable Autoclavable
Material Polypropylene
Color Natural
Quantity 1000/Pk.
For Use With (Equipment) Globe Scientific sample tubes with external threads (21-200-102)
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.