missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SD 2590 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
SD 2590 hydrochloride is a potent MMP inhibitor (IC50 values are <0.1, <0.1, 0.18, 1.7, 13 and 28.7 nM for MMP-2, -13, -9, -8, -14 and -3 respectively). Exhibits 100,000-fold selectivity for MMP-2 and -13 over MMP-1. Reduces left ventricular dilation in a myocardial infarction rat model. Orally bioavailable.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | SD 2590 hydrochloride |
| Purity | >98% |
| Molecular Weight (g/mol) | 554.96 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction