Learn More
Trajan Scientific and Medical™ Seal-seat Removal Tool
Product Code. 15279153
Change view
Click to view available options
For Use With (Equipment):
For Micro Control Valve
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15279153 For Micro Control Valve Each
1 options
Product Code. 15279153 Supplier Trajan Scientific and Medical™ Supplier No. 1236101
Only null left
Add to basket
Add to basket
For use with Micro Control Valves. Trajan Scientific and Medical Seal-seat Removal Tool used with Micro Control Valves provides precision metering and on-off control of gas flows
  • Ideal for chromatography applications
  • May be used at elevated temperatures from high vacuum to medium pressure
  • Vacuum rated to a leak rate of 1E-05 cc/sec of helium
Specifications
Type Seal Seat Removal Tool for Micro Control Valve
For Use With (Equipment) For Micro Control Valve