Learn More
Prestige Medical™ Sealing Gasket
Product Code. 13466429
Change view
Click to view available options
Quantity:
Pk of 4
Unit Size:
Pack of 4
Product Code. Quantity Unit Size
13466429 Pk of 4 Pack of 4
1 options
Product Code. 13466429 Supplier Prestige Medical™ Supplier No. 219748
Estimated Shipment: 23-11-2026
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Sealing gaskets For Classic models with a sterlising temperature of 121°C/126°C, green.
  • An important safety feature of your autoclave
  • Correct maintenance and replacement of your sealing gasket will help prevent unnecessary breakdowns and expensive down time
Specifications
Quantity Pk of 4
Type Sealing Gasket