missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Sealing ring for Dessicator

Product Code. 10568471 Shop All Brand Products
Change view
Click to view available options
Description:
Nominal size 200 mm
Nominal size 250 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Unit Size
10568471 Nominal size 250 mm Each
10017780 Nominal size 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10568471 Supplier BRAND™ Supplier No. 65824
  • ONLINE EXCLUSIVE PRICE
    20.25 €
    15.38 € / Pack of 1
    Save 4.87 €
    24% Off
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Provides seal for dessicator applications

  • CR construction
  • For use with Dessicators
TRUSTED_SUSTAINABILITY

Specifications

Description Nominal size 250 mm
Material Chloroprene Rubber
For Use With (Equipment) For Desiccators
Product Type Desiccator Sealing Ring
Quantity 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.