Learn More
DWK Life Sciences Sealing ring, for polished spherical joints (silicone + FEP)
Shop All DWK Life Sciences ProductsDescription
Non-sanded DURAN™ spherical ground joints only seal via the sealing ring. FEP-coated silicone rings are used where high chemical resistance is required.
- Due to the material properties, it can be used for work with high demands on the chemical resistance of the sealing ring.
- Silicone and FEP sealing ring
Specifications
Specifications
| Description | 41 NS |
| Autoclavable | Not Autoclavable |
| Material | Silicone, FEP |
| For Use With (Application) | Sealing of non-sanded spherical ground joints |
| Packaging Quantity | 10/Pk. |