missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ Seamless Knit Nylon Gloves with Polyurethane Coated Fingertips

Product Code. 17851201
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
10 pairs
Product Code. Size unitSize
17851201 Small 10 pairs
17717627 Small 10 pairs
17707627 Medium 10 pairs
17861201 Large 10 pairs
17871201 X-Large 10 pairs
17727627 X-Large 10 pairs
6 options
This item is not returnable. View return policy

Product Code. 17851201

Brand: SHOWA™ BO60006S

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

General purpose gloves feature a snug fit and polyurethane coated fingertips for sensitivity and control. Permeable construction allows the hands to breathe and maintain temperature control.

  • Polyurethane fingertips provide touch sensitivity
  • Coating is permeable, allowing the hands to breathe
  • Deformation-resistant glove shape is maintained for an extended period of time
  • No fingerprints are left on clear, clean objects
  • Snug fit maximizes tactile feedback
  • Designed for precision applications

Specifications

Coated Area Fingertips
Construction Seamless Knit
Cuff Material Elastic
Coating Material Polyurethane
Color White
Exterior Finish No Texture
Hand Specific
Material Nylon
Product Type Seamless Knit Gloves
Size Small
EN388 Standard 0131X
For Use With (Application) Electronic Components, Quality Control, and Integrated Circuits
Glove Cuff Color Natural
Glove Cuff Style Knit
Series BO600
Yarn Gauge 13
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.