missing translation for 'onlineSavingsMsg'
Learn More

sec-Butanol, 99+%, Extra Dry, AcroSeal™

Product Code. 10653605 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
1 L
100mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10653605 1 L AcroSeal™ Glass Bottle 1 L
10537264 100 mL AcroSeal™ Glass Bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10653605 Supplier Thermo Scientific Acros Supplier No. 610260010
  • ONLINE EXCLUSIVE PRICE
    212.00 €
    205.00 € / 1 L
    Save 7.00 €
    3% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78-92-2 | C4H10O | 74.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-92-2
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004569
InChI Key BTANRVKWQNVYAZ-UHFFFAOYNA-N
Synonym 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
PubChem CID 6568
ChEBI CHEBI:35687
IUPAC Name butan-2-ol
SMILES CCC(C)O

Specifications

Density 0.81g/mL
Quantity 1 L
Identification Pass Test
Packaging AcroSeal™ Glass Bottle
Assay Percent Range 99%
Linear Formula C2H5CH(OH)CH3
Water 50ppm max.
Beilstein 01, 371
Merck Index 15, 1543
Formula Weight 74.12
Percent Purity ≥99%
Grade Extra Dry
Chemical Name or Material sec-Butanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • sec-Butanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Butanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.