missing translation for 'onlineSavingsMsg'
Learn More

sec-Butanol, 99%, Extra Pure

Product Code. 10448630 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic Drum
Unit Size:
1 L
10 L
2.5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10448630 1 L Glass Bottle 1 L
10765361 2.5 L Glass Bottle 2.5 L
10512744 10 L Plastic Drum 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10448630 Supplier Thermo Scientific Acros Supplier No. 107700010
  • ONLINE EXCLUSIVE PRICE
    37.10 €
    34.10 € / 1 L
    Save 3.00 €
    8% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78-92-2 | C4H10O | 74.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-92-2
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004569
InChI Key BTANRVKWQNVYAZ-UHFFFAOYNA-N
Synonym 2-butanol, sec-butanol, sec-butyl alcohol, 2-hydroxybutane, 2-butyl alcohol, s-butyl alcohol, butylene hydrate, s-butanol, 1-methyl-1-propanol, 1-methylpropyl alcohol
PubChem CID 6568
ChEBI CHEBI:35687
IUPAC Name butan-2-ol
SMILES CCC(C)O

Specifications

Density 0.8100g/mL
Quantity 1 L
Infrared Spectrum Conforms to Ref Report
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3960 to 1.3980 (20°C, 589nm)
Residue after Evaporation 0.005% max.
Assay Percent Range 99%
Linear Formula C2H5CH(OH)CH3
Water 0.5% max.
Beilstein 01, 371
Merck Index 15, 1543
Formula Weight 74.12
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material sec-Butanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • sec-Butanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.