missing translation for 'onlineSavingsMsg'
Learn More

Sec-Butyl Alcohol, Certified AR for Analysis, Fisher Chemical™

Product Code. 10751394 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber glass bottle
Plastic Coated Glass bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10751394 1 L Amber glass bottle 1 L
10508420 2.5 L Plastic Coated Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10751394 Supplier Fisher Chemical Supplier No. B/5210/15
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 78-92-2
Molecular Formula C4H10O
MDL Number 4569

Specifications

Melting Point -115°C
Color Colorless
Boiling Point 99°C
Packaging Amber glass bottle
Quantity 1 L
Vapor Density 2.6
Formula Weight 74.12g/mol
Vapor Pressure 17mbar at 20°C
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • sec-Butyl alcohol
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H360-May damage fertility or the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.